missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgM κ, Mouse, FITC, Clone: G155-228, Isotype Control, BD
364.00 CHF - 375.00 CHF
Spezifikation
Klon | G155-228 |
---|---|
Anwendungen | Flow Cytometry, Immunofluorescence, Western Blot |
Konjugat | FITC |
Wirtsspezies | Mouse |
Zielspezies | Non-human Primate |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15847238
|
BD Biosciences
553474 |
0.25 mg |
375.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15826658
|
BD Biosciences
551448 |
50 Tests |
364.00 CHF
1 Stück |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The G155-228 antibody is specific for the hapten trinitrophenol (TNP), a hapten not expressed on non-human primate cells or non-human primate cell lines. In the absence of specific binding, this immunoglobulin may bind non-specifically to Fc receptors.
Host Species: Mouse
Clone: G155-228
Isotype: IgM, κ
Species Reactivity: Non-human primate
Immunogen: TNP-Keyhole Limpet Hemocyanin
Flow Cytometry, Immunofluorescence, Intracellular Staining
Spezifikation
G155-228 | |
FITC | |
Non-human Primate | |
IgM κ | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
Flow Cytometry, Immunofluorescence, Western Blot | |
Mouse | |
TNP-keyhole limpet hemocyanin | |
Affinity Purified |