Learn More
anti-IgG2, κ, Clone: B81-3, BD
Marke: BD Biosciences 559339
Additional Details : Gewicht : 0.01000kg
Beschreibung
The B81-3 antibody is specific for keyhole limpet hemocyanin. This antibody is routinely tested by LAL assay for endotoxin levels. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Blocking Assay, Co-Stimulation, Cytotoxicity Assay, Isotype Control
Spezifikation
Blocking Assay, Functional Assay, Cytotoxicity Assay | |
1mg/mL | |
Hamster | |
0.5 mg | |
IgG2 κ |
B81-3 | |
Unconjugated | |
Affinity Purified | |
Undiluted at 4°C |