Learn More
anti-IgG1 κ, Mouse, APC, Clone: MOPC-21, Isotype Control, BD
Marke: BD Biosciences 554681
578.87 CHF gültig bis 2025-12-31
Benutzen Sie den Promotions-Code "25339" um Ihren Promotions-Preis zu erhalten.
Beschreibung
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Non-human primate
Flow Cytometry, Intracellular Staining, Isotype Control

Spezifikation
Flow Cytometry, Western Blot | |
0.2mg/mL | |
Mouse | |
0.1 mg | |
Undiluted at 4°C |
MOPC-21 | |
APC | |
Affinity Purified | |
Non-human Primate | |
IgG1 κ |
Produktvorschläge
Customers who viewed this item also viewed.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.