missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1 κ, Mouse, APC, Clone: MOPC-21, Isotype Control, BD
368.00 CHF - 910.00 CHF
Spezifikation
Klon | MOPC-21 |
---|---|
Konzentration | 0.2mg/mL |
Anwendungen | Flow Cytometry, Western Blot |
Konjugat | APC |
Wirtsspezies | Mouse |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15827838
|
BD Biosciences
554681 |
0.1 mg |
910.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15856408
|
BD Biosciences
550854 |
50 Tests |
368.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Non-human primate
Flow Cytometry, Intracellular Staining, Isotype Control
Spezifikation
MOPC-21 | |
Flow Cytometry, Western Blot | |
Mouse | |
IgG1 κ | |
Undiluted at 4°C |
0.2mg/mL | |
APC | |
Non-human Primate | |
Affinity Purified |