Learn More
Disabled-2/p96 Mouse, Unlabeled, Clone: 52, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 610465
Beschreibung
CSF-1 is a growth factor that stimulates the growth and differentiation of immature lymphocytes and is required for the survival of mononuclear phagocytes. Binding of CSF-1 induces dimerization and autophosphorylation of its receptor. This results in the activation of several signal transduction pathways. A unique 96 kDa protein is a component in the CSF-1 signal transduction cascade. p96 is phosphorylated on serine following mitogenic stimulation of a mouse macrophage cell line. p96 contains three potential C-terminal ERK kinase phosphorylation sites, as well as several proline-rich sequences that are potential binding sites for SH3-containing proteins. Structural similarities have been found between p96 and Dab, a product of the Drosophila disabled gene, and p96 was also identified as Disabled-2 (Dab-2) and as differentially expressed in ovarian β receptor to the Smad family of transcription factors. Thus, Dab-2/p96 is an important adaptor molecule in growth factor signaling pathways.
Spezifikation
| Disabled-2/p96 | |
| Monoclonal | |
| 250μg/mL | |
| Disabled-2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 52 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.