missing translation for 'onlineSavingsMsg'
Learn More

CD95 Mouse, Unlabeled, Clone: 13, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610198

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15835329

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.


Beschreibung

Beschreibung

CD95 is a member of a family of cell surface receptors that includes tumor necrosis factor receptor (TNF-R), nerve growth factor receptor (NGF-R), CD40, CD27, CD30, and 4-1BB. Both murine and human Fas genes have been cloned and reportedly share 60% similarity in their amino acid sequences. CD95 (Fas) is a cell surface apoptosis-signaling molecule that is widely expressed in sites such as thymus, liver, heart, and ovary. Abnormalities in the Fas gene correlate with autoimmune features in mice and with unusually high levels of lymphocyte apoptosis in HIV-infected humans. Genetic studies have localized the Fas gene near the lpr (lymphoproliferation disease) locus on mouse chromosome 19 and further characterization reportedly has revealed that lpr is a mutation affecting the function of the Fas gene.

Spezifikation

Spezifikation

CD95
Monoclonal
250μg/mL
Fas, APO-1
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
13
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.