missing translation for 'onlineSavingsMsg'
Learn More

CD95 Mouse, Unlabeled, Clone: 13, BD

Mouse Monoclonal Antibody

679.00 CHF

Spezifikation

Antigen CD95
Klon 13
Konzentration 250μg/mL
Anwendungen Western Blot
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 1
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15835329
Unterlagen ansehen Aktionsdetails
BD Biosciences
610198
150 μg Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
N/A Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Beschreibung

Beschreibung

CD95 is a member of a family of cell surface receptors that includes tumor necrosis factor receptor (TNF-R), nerve growth factor receptor (NGF-R), CD40, CD27, CD30, and 4-1BB. Both murine and human Fas genes have been cloned and reportedly share 60% similarity in their amino acid sequences. CD95 (Fas) is a cell surface apoptosis-signaling molecule that is widely expressed in sites such as thymus, liver, heart, and ovary. Abnormalities in the Fas gene correlate with autoimmune features in mice and with unusually high levels of lymphocyte apoptosis in HIV-infected humans. Genetic studies have localized the Fas gene near the lpr (lymphoproliferation disease) locus on mouse chromosome 19 and further characterization reportedly has revealed that lpr is a mutation affecting the function of the Fas gene.

Spezifikation

Spezifikation

CD95
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
Fas, APO-1
13
Western Blot
Unconjugated
RUO
Canine, Chicken, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

For Research Use Only.