Learn More
CD3 Molecular Complex Rat anti-Mouse, FITC, Clone: 17A2, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 561798
147.66 CHF gültig bis 2025-12-31
Benutzen Sie den Promotions-Code "25339" um Ihren Promotions-Preis zu erhalten.
Beschreibung
The 17A2 monoclonal antibody specifically binds to the T-cell receptor-associated CD3 complex that is expressed on many thymocytes and mature T lymphocytes. Plate-bound 17A2 antibody has been reported to induce IL-2 production by cultured T cells in the absence of accessory cells. The binding of the 17A2 antibody to T cells can be blocked by the anti-CD3e mAb 145-2C11. This suggests that the 17A2 antibody recognizes an epitope of the CD3 epsilon chain. In vivo treatment with 17A2 antibody has been reported to partially deplete T lymphocytes and temporarily down-modulates CD3 expression on T cells.

Spezifikation
CD3 Molecular Complex | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
γδ TCR-positive T-T hybridoma D1 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
17A2 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.