Learn More
CD29 Mouse anti-Human, Brilliant Violet 510, Clone: MAR4, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 563514
Additional Details : Gewicht : 0.01000kg
Beschreibung
The MAR4 monoclonal antibody specifically binds to CD29. CD29 is a 130 kDa integrin β1 subunit that is expressed as a heterodimeric complex with one of six distinct α subunits, comprising the very late activation antigen (VLA) subfamily of adhesion receptors. The β1 subunit has a broad tissue distribution; it is expressed on lymphocytes, monocytes and weakly on granulocytes, but not on erythrocytes. These receptors are involved in a variety of cell-cell and cell-matrix interactions.
Spezifikation
CD29 | |
Monoclonal | |
Brilliant Violet 510 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Purified Human α5β1 Fibronectin Receptor | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
Mar4 | |
ITGB1; Integrin beta-1; Integrin β1; FNRB; GPIIA; ITB1; MDF2; MSK12; VLA-4β | |
Mouse | |
Affinity Purified | |
RUO | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
For Research Use Only.