missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Anisomycin
GREENER_CHOICE

JNK, SAPK and p38 activator

177.00 CHF - 553.00 CHF

Chemische Identifikatoren

CAS 22862-76-6
Summenformel C14H19NO4
Molekulargewicht 265.31
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
18712664
Unterlagen ansehen
Tocris Bioscience™
1290/10
10 mg
177.00 CHF
10 Milligramm
Nur noch null auf Lager
Verfügbar ab: 21-07-2025
Zum Warenkorb hinzufügen
 
18712504
GREENER_CHOICE
Unterlagen ansehen
Tocris Bioscience™
1290/50
50 mg
553.00 CHF
50 Milligramm
Only null left
Verfügbar ab: 21-07-2025
Add to basket
 
Description

Description

Protein synthesis inhibitor (blocks translation). Potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. Acts as a potent signaling agonist to selectively elicit homologous desensitization of immediate early gene induction (c-fos, fosB, c-jun, junB and junD).
Specifications

Chemical Identifiers

22862-76-6
C14H19NO4
265.31

Specifications

Anisomycin
C14H19NO4
Anisomycin
265.31
22862-76-6
265.31
>98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at +4°C

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tocris Bioscience™ Anisomycin

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.