missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphiphysin/AMPH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
418.00 CHF - 582.00 CHF
Spezifikation
| Antigen | Amphiphysin/AMPH |
|---|---|
| Verdünnung | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Anwendungen | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18438261
|
Novus Biologicals
NBP1-87561 |
0.1 mL |
582.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18435302
|
Novus Biologicals
NBP1-87561-25ul |
25 μL |
418.00 CHF
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Amphiphysin/AMPH Polyclonal antibody specifically detects Amphiphysin/AMPH in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Spezifikation
| Amphiphysin/AMPH | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Immune System Diseases, Immunology, Tumor Suppressors | |
| PBS (pH 7.2) and 40% Glycerol | |
| 273 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| AMPH1, amphiphysin, amphiphysin (Stiff-Man syndrome with breast cancer 128kDa autoantigen), amphiphysin (Stiff-Mann syndrome with breast cancer 128kD autoantigen), amphiphysin I | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EKVIPSVVIEPASNHEEEGENEITIGAEPKETTEDAAPPGPTSETPELATEQKPIQDPQPTPSAPAMGAADQLASAREA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts