missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALPK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19107-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Unlike most eukaryotic kinases, alpha kinases, such as LAK, recognize phosphorylation sites in which the surrounding peptides have an alpha-helical conformation.Spezifikation
ALPK1 | |
Polyclonal | |
Unconjugated | |
ALPK1 | |
8430410J10Rik, alpha kinase 1, Alpha protein kinase 1, ALPK1, Chromosome 4 kinase, KIAA1527, LAK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
310879, 71481, 80216 | |
Antibody | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96QP1, Q9CXB8 | |
Alpk1 | |
ALPK1 Fusion Protein Ag5320 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Liquid |