Learn More
Novus Biologicals™ ALK-1 Antibody (ascrinvacumab) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Marke: Novus Biologicals™ NBP3-28297-1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ALK-1 Monoclonal antibody specifically detects ALK-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spezifikation
| ALK-1 | |
| Monoclonal | |
| Unconjugated | |
| P37023 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| ascrinvacumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I | |
| ACVRL1/ ALK-1 | |
| 1 mg | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 94 | |
| Human | |
| Purified |