Learn More
Invitrogen™ Human AKT1 (PKB alpha), His Tag Recombinant Protein
Human AKT1 (PKB alpha), His Tag Recombinant Protein
Marke: Invitrogen™ PR3878D
Additional Details : Gewicht : 0.01000kg
Beschreibung
The activated AKT1 (PKB alpha) enzyme inhibits apoptosis and stimulates cell cycle progression by phosphorylating numerous targets in various cell types, including cancer cells.
Spezifikation
NP_005154 | |
59.1 kDa | |
AKT1 (PKB alpha) | |
Human | |
His Tag | |
Ser⁄Thr Kinases: ACG Group | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
100 μg | |
AKT Kinase Family | |
Recombinant | |
Storage Buffer: 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.