Learn More
AHR Antibody (RPT9) - BSA Free, Novus Biologicals™
Artikelnummer. 18720414 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18720414 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18720414 Lieferant Novus Biologicals Lieferanten-Nr. NB300515
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Mouse Monoclonal Antibody has been used in 7 publications
AHR Monoclonal specifically detects AHR in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Gel Super Shift Assays.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen AHR
Anwendungen Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Klassifikation Monoclonal
Klon RPT9
Konzentration 1 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 1:500 - 1:2000, Flow Cytometry 1:50, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:100
Zusammensetzung No buffer with 0.05% Sodium Azide
Gen-Zugriffsnummer P30561
Gen-Alias Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76
Gensymbole AHR
Wirtsspezies Mouse
Immunogen Synthetic peptide corresponding to residues R(12) K R R K P(17) V(22) K P I P A E G I K(31) of mouse AhR.
Reinigungsverfahren Protein A purified
Menge 100 μL
Regulatorischer Status RUO
Forschungsgebiet Apoptosis, Cancer, Hypoxia, Innate Immunity, Transcription Factors and Regulators
Primär oder sekundär Primary
Gen-ID (Entrez) 196
Testspezifität Detects the Aryl Hydrocarbon Receptor (AHR).
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3 κ