missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12825-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.Spezifikation
AF9 | |
Polyclonal | |
Unconjugated | |
MLLT3 | |
AF9, FLJ2035, MLLT3, Protein AF 9, YEATS3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114510, 4300, 70122 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2AM29, P42568 | |
MLLT3 | |
AF9 Fusion Protein Ag3528 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |