missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Specifications
Antigen | AF9 |
---|---|
Konzentration | 0.13 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
16850744
|
Proteintech
12825-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840744
|
Proteintech
12825-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Description
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.Specifications
AF9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A2AM29, P42568 | |
114510, 4300, 70122 | |
AF9 Fusion Protein Ag3528 | |
Primary | |
-20°C | |
MLLT3 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AF9, FLJ2035, MLLT3, Protein AF 9, YEATS3 | |
MLLT3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |