missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADRBK2 (GRK3) Recombinant Human Protein
ADRBK2, or adrenergic beta receptor kinase 2 is a serine/threonine kinase.
Marke: Invitrogen™ PV3974
Additional Details : Gewicht : 0.00900kg
Beschreibung
ADRBK2 is thought to act as a regulator of receptor function.
Spezifikation
NP_005151.2 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
ADRBK2 (GRK3) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
107.2kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
GRK Kinase Family | |
Recombinant | |
Baculovirus |