missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADRA1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
129.00 CHF - 349.00 CHF
Spezifikation
Antigen | ADRA1B |
---|---|
Konzentration | 0.24 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin) |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839254
|
Proteintech
19776-1-AP-20UL |
20 μL |
129.00 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829254
|
Proteintech
19776-1-AP-150UL |
150 μL |
349.00 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This antibody is specific to ADRA1B.
The alpha-1b adrenoceptor is an Adrenergic Receptor that causes contraction of smooth muscle cells and thereby controls vascular tone, blood pressure, and accelerates the development of atherosclerosis. Decreased alpha-1b adrenoceptor is associated with benign prostatic hyperplasia (BPH). This adrenergic receptor also acts as a proto-oncogene when transfected into cell lines where constitutive activation of the receptor induces neoplastic transformation. The alpha-1b adrenoceptor has been documented in adrenal, aorta, blood, various brain regions, eye, heart, kidney, liver, prostate, spinal cord, and spleen. ESTs have been isolated from kidney (normal and cancer) and lung carcinoma libraries.Spezifikation
ADRA1B | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P15823, P35368, P97717 | |
11548, 147, 24173 | |
Peptide | |
Primary | |
-20°C | |
ADRA1B |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ADRA1, ADRA1B, Alpha 1B adrenergic receptor, Alpha 1B adrenoceptor, Alpha 1B adrenoreceptor, ALPHA1BAR | |
Adra1b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |