missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADRA1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19776-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody is specific to ADRA1B.
The alpha-1b adrenoceptor is an Adrenergic Receptor that causes contraction of smooth muscle cells and thereby controls vascular tone, blood pressure, and accelerates the development of atherosclerosis. Decreased alpha-1b adrenoceptor is associated with benign prostatic hyperplasia (BPH). This adrenergic receptor also acts as a proto-oncogene when transfected into cell lines where constitutive activation of the receptor induces neoplastic transformation. The alpha-1b adrenoceptor has been documented in adrenal, aorta, blood, various brain regions, eye, heart, kidney, liver, prostate, spinal cord, and spleen. ESTs have been isolated from kidney (normal and cancer) and lung carcinoma libraries.Spezifikation
ADRA1B | |
Polyclonal | |
Unconjugated | |
ADRA1B | |
ADRA1, ADRA1B, Alpha 1B adrenergic receptor, Alpha 1B adrenoceptor, Alpha 1B adrenoreceptor, ALPHA1BAR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11548, 147, 24173 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P15823, P35368, P97717 | |
Adra1b | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |