Learn More
ADO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16479-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Human thiol dioxygenases include cysteine dioxygenase (CDO; MIM 603943) and cysteamine (2-amino ethane thiol) dioxygenase (ADO; EC 1.13.11.19). CDO adds 2 oxygen atoms to free cysteine, whereas ADO adds 2 oxygen atoms to free cysteamine to form hypotaurine.Spezifikation
| ADO | |
| Polyclonal | |
| Unconjugated | |
| ADO | |
| 2 aminoethanethiol dioxygenase, ADO, C10orf22, Cysteamine dioxygenase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 211488, 309732, 84890 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PDY2, Q96SZ5 | |
| Ado | |
| ADO Fusion Protein Ag9598 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |