missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin receptor 1 Mouse anti-Human, Clone: 1F12B3, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66048-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody was raised against 1-72 aa of human AdipoR1.
The adiponectin receptors, ADIPOR1 and ADIPOR2, serve as receptors for globular and full-length adiponectin and mediate increased AMPK and PPAR-alpha ligand activities, as well as fatty acid oxidation and glucose uptake by adiponectin.Spezifikation
Adiponectin receptor 1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96A54 | |
ADIPOR1 | |
Adiponectin receptor 1 Fusion Protein Ag18187 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
1F12B3 | |
Unconjugated | |
ADIPOR1 | |
ACDCR1, Adiponectin receptor 1, Adiponectin receptor protein 1, ADIPOR1, CGI 45, CGI45, PAQR1, TESBP1A | |
Mouse | |
Protein G | |
RUO | |
51094 | |
-20°C | |
Liquid |