missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin receptor 1 Mouse anti-Human, Clone: 1F12B3, Proteintech
Mouse Monoclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | Adiponectin receptor 1 |
---|---|
Klon | 1F12B3 |
Konzentration | 1 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16848665
|
Proteintech
66619-1-IG-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16838665
|
Proteintech
66048-1-IG-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This antibody was raised against 1-72 aa of human AdipoR1.
The adiponectin receptors, ADIPOR1 and ADIPOR2, serve as receptors for globular and full-length adiponectin and mediate increased AMPK and PPAR-alpha ligand activities, as well as fatty acid oxidation and glucose uptake by adiponectin.Spezifikation
Adiponectin receptor 1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACDCR1, Adiponectin receptor 1, Adiponectin receptor protein 1, ADIPOR1, CGI 45, CGI45, PAQR1, TESBP1A | |
ADIPOR1 | |
IgG1 | |
Protein G | |
Antibody |
1F12B3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q96A54 | |
51094 | |
Adiponectin receptor 1 Fusion Protein Ag18187 | |
Primary | |
-20°C | |
ADIPOR1 |