Learn More
Acebutolol hydrochloride, Thermo Scientific™
Cardioselective beta-adrenergic blocker
62.79 CHF - 228.72 CHF
Chemische Identifikatoren
| CAS | 34381-68-5 |
|---|---|
| Summenformel | C18H29ClN2O4 |
| Molekulargewicht (g/mol) | 372.89 |
| MDL-Nummer | MFCD00078860 |
| InChI-Schlüssel | KTUFKADDDORSSI-UHFFFAOYNA-N |
| Synonym | acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn |
| PubChem CID | 441307 |
| ChEBI | CHEBI:2380 |
| IUPAC-Name | hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride |
| SMILES | [H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15445959
|
Thermo Scientific Alfa Aesar
J63790.03 |
1g |
64.45 CHF 62.79 CHF / 1 Gramm Sparen 1.66 CHF 2% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
15455959
|
Thermo Scientific Alfa Aesar
J63790.06 |
5g |
134.00 CHF 129.90 CHF / 5 Gramm Sparen 4.10 CHF 3% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
15465959
|
Thermo Scientific Alfa Aesar
J63790.09 |
10g |
236.00 CHF 228.72 CHF / 10 Gramm Sparen 7.28 CHF 3% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
| 34381-68-5 | |
| 372.89 | |
| KTUFKADDDORSSI-UHFFFAOYNA-N | |
| 441307 | |
| hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride |
| C18H29ClN2O4 | |
| MFCD00078860 | |
| acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn | |
| CHEBI:2380 | |
| [H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O |
Spezifikation
| Acebutolol hydrochloride | |
| C18H29ClN2O4 | |
| 14,19 | |
| KTUFKADDDORSSI-UHFFFAOYNA-N | |
| hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride | |
| 441307 | |
| 372.89 |
| 34381-68-5 | |
| MFCD00078860 | |
| acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn | |
| [H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O | |
| 372.89 | |
| CHEBI:2380 |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 251-980-3
missing translation for 'rtecsNumber' : ES5235000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.