Learn More
Acebutolol hydrochloride, Thermo Scientific™
Cardioselective beta-adrenergic blocker
Marke: Thermo Scientific Alfa Aesar J63790.06
| Menge | 5g |
|---|
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
| 34381-68-5 | |
| 372.89 | |
| KTUFKADDDORSSI-UHFFFAOYNA-N | |
| 441307 | |
| hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride |
| C18H29ClN2O4 | |
| MFCD00078860 | |
| acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn | |
| CHEBI:2380 | |
| [H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O |
Spezifikation
| Acebutolol hydrochloride | |
| C18H29ClN2O4 | |
| 5g | |
| acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn | |
| [H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O | |
| 372.89 | |
| CHEBI:2380 |
| 34381-68-5 | |
| MFCD00078860 | |
| 14,19 | |
| KTUFKADDDORSSI-UHFFFAOYNA-N | |
| hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride | |
| 441307 | |
| 372.89 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.