missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ 1800 mm ReVo Class II, Type A2 Microbiological Safety Cabinet
ReVo™ Class II, Type A2 Microbiological Safety Cabinets provide personnel, product and environmental protection from biohazardous agents and hazardous particulates.
16710.00 CHF - 17585.00 CHF
Spezifikation
Zertifizierung/Konformität | EN 12469 Certified by TÜV |
---|---|
Kammermaterial | Stainless Steel (304) |
Produkttyp | Class II Biosafety Cabinet |
Geräuschpegel | <55 dBA |
Steckdosen | 2 |
Produktcode | Marke | Anschlüsse | Enthält | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Anschlüsse | Enthält | Preis | Menge & Verfügbarkeit | ||||
17890754
|
Labconco™
3R2610031 |
Schuko Plug | Base Stand Not Included |
16710.00 CHF
1 Einheit |
Verfügbar ab: 04-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
17880754
|
Labconco™
3R2610051 |
UK Plug | Base Stand Not Included |
16710.00 CHF
1 Einheit |
Verfügbar ab: 04-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
17880744
|
Labconco™
3R2610131 |
Schuko Plug | Base Stand |
17585.00 CHF
1 Einheit |
Verfügbar ab: 04-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
17810764
|
Labconco™
3R2610151 |
UK Plug | Base Stand |
17585.00 CHF
1 Einheit |
Verfügbar ab: 04-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
Beschreibung
- Provide personnel, product and environmental protection from biohazardous agents and hazardous particulates.
- Other appropriate applications include work with antineoplastic drugs, genetic material, and additional substances that generate hazardous airborne particulates..
- All models are EN 12469 compliant with additional certification by TÜV Nord.
- These biological safety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection.
- When canopy-connected, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.
- During operation, room air is drawn into the inlet grill.
- The air in the plenum beneath the work surface is a mixture of unfiltered room air and air that has just passed through the work area.
- This contaminated air is drawn by the blower through the back plenum of the cabinet, where approximately 70% of the air is recirculated through the supply HEPA filter and back over the work area.
- The balance of the contaminated air is discharged to the environment after passing through the exhaust HEPA filter.
Spezifikation
EN 12469 Certified by TÜV | |
Class II Biosafety Cabinet | |
2 | |
200 mm | |
7.87 in. | |
230 V |
Stainless Steel (304) | |
<55 dBA | |
90 W | |
HEPA Exhaust Filter | |
797 x 1941 x 1350 mm |