missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ 1800 mm ReVo Class II, Type A2 Microbiological Safety Cabinet

ReVo™ Class II, Type A2 Microbiological Safety Cabinets provide personnel, product and environmental protection from biohazardous agents and hazardous particulates.

Marke:  Labconco™ 3R2610031

Additional Details : Gewicht : 262.00000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 17890754

  • 16710.00 CHF / 1 Einheit
Verfügbar ab: 18-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Provide personnel, product and environmental protection from biohazardous agents and hazardous particulates.
  • Other appropriate applications include work with antineoplastic drugs, genetic material, and additional substances that generate hazardous airborne particulates..
  • All models are EN 12469 compliant with additional certification by TÜV Nord.
  • These biological safety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection.
  • When canopy-connected, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.
  • During operation, room air is drawn into the inlet grill.
  • The air in the plenum beneath the work surface is a mixture of unfiltered room air and air that has just passed through the work area.
  • This contaminated air is drawn by the blower through the back plenum of the cabinet, where approximately 70% of the air is recirculated through the supply HEPA filter and back over the work area.
  • The balance of the contaminated air is discharged to the environment after passing through the exhaust HEPA filter.
Spezifikation

Spezifikation

EN 12469 Certified by TÜV
Class II Biosafety Cabinet
2
200 mm
Base Stand Not Included
7.87 in.
262.0 kg
Stainless Steel (304)
<55 dBA
90 W
HEPA Exhaust Filter
Schuko Plug
797 x 1941 x 1350 mm
230 V
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote