missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals

Product Code. 11308976
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308976 25 g 25g
11398966 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308976 Supplier Thermo Scientific Alfa Aesar Supplier No. L20158.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 618 41 11 for assistance
View alternative products

This item is not returnable. View return policy

It is used in the stereoselective synthesis of four stereoisomers of β-Methoxytyrosine, a component of callipeltin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the stereoselective synthesis of four stereoisomers of β-Methoxytyrosine, a component of callipeltin A.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60876-70-2
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.117
MDL Number MFCD00792676
InChI Key QIWQHUCUWNGYDZ-UHFFFAOYSA-N
Synonym 1-bromo-4-tert-butoxy benzene, 1-bromo-4-tert-butoxybenzene, 4-tert-butoxybromobenzene, 4-bromophenyl tert-butyl ether, 1-bromo-4-tert-butoxy-benzene, benzene, 1-bromo-4-1,1-dimethylethoxy, tert-butyl-4-bromophenyl ether, 1-bromo-4-2-methylpropan-2-yl oxy benzene, 1-tert-butoxy-4-bromobenzene, bromobutoxybenzene
PubChem CID 2763959
IUPAC Name 1-bromo-4-[(2-methylpropan-2-yl)oxy]benzene
SMILES CC(C)(C)OC1=CC=C(C=C1)Br

Specifications

Boiling Point 63°C to 65°C (0.4 mmHg)
Quantity 25 g
Beilstein 2516411
Formula Weight 229.12
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-tert-butoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.