CAS RN 2379-55-7
CAS RN 2379-55-7
IUPAC Name:
2,3-dimethylquinoxaline
Synonyme:
quinoxaline, 2,3-dimethyl-
Molekulargewicht (g/mol):
158.2
Summenformel:
C10H10N2
InChi Key:
FKHNZQFCDGOQGV-UHFFFAOYSA-N
SMILES:
CC1=C(C)N=C2C=CC=CC2=N1
1
–
2
von
2
Ergebnisse
1
2,3-Dimethylchinoxalin, 97 %, Thermo Scientific Chemicals
CAS: 2379-55-7 Summenformel: C10H10N2 Molekulargewicht (g/mol): 158.204 MDL-Nummer: MFCD00006728 InChI-Schlüssel: FKHNZQFCDGOQGV-UHFFFAOYSA-N PubChem CID: 16925 IUPAC-Name: 2,3-Dimethylchinoxalin SMILES: CC1=NC2=CC=CC=C2N=C1C
2
2,3-Dimethylchinoxalin 97 %, Thermo Scientific Chemicals
CAS: 2379-55-7 MDL-Nummer: MFCD00006728 InChI-Schlüssel: FKHNZQFCDGOQGV-UHFFFAOYSA-N PubChem CID: 16925 IUPAC-Name: 2,3-Dimethylchinoxalin SMILES: CC1=NC2=CC=CC=C2N=C1C