CAS RN 850568-71-7
IUPAC Name:
(quinolin-3-yl)boronic acid hydrochloride
Synonyme:
quinolin-3-ylboronic acid hydrochloride
Molekulargewicht (g/mol):
209.44
Summenformel:
C9H9BClNO2
InChi Key:
BQFQABXCFQWPGH-UHFFFAOYSA-N
SMILES:
Cl.OB(O)C1=CN=C2C=CC=CC2=C1
1
–
1
von
1
Ergebnisse
1
Chinolin-3-Boronsäurehydrochlorid, 97 %, Thermo Scientific™
CAS: 850568-71-7 Summenformel: C9H9BClNO2 Molekulargewicht (g/mol): 209.44 MDL-Nummer: MFCD06659836 InChI-Schlüssel: BQFQABXCFQWPGH-UHFFFAOYSA-N Synonym: quinolin-3-ylboronic acid hydrochloride,quinoline-3-boronic acid hydrochloride,3-quinolineboronic acid hydrochloride,quinoline-3-boronic acid hcl,quinoline-3-boronic acid, hcl,3-quinolineboronic acid, hcl,quinoline-3-boronic acid,hydrochloride,pubchem5872,3-quinolinylboronic acid hydrochloride PubChem CID: 44118768 IUPAC-Name: Chinolin-3-ylboronsäure;hydrochlorid SMILES: Cl.OB(O)C1=CN=C2C=CC=CC2=C1