CAS RN 19883-41-1
CAS RN 19883-41-1
IUPAC Name:
hydrogen methyl 2-(2-aminophenyl)acetate chloride
Synonyme:
hydrogen methyl 2-(2-aminophenyl)acetate chloride
Molekulargewicht (g/mol):
201.65
Summenformel:
C9H12ClNO2
InChi Key:
OZMUKUBEEKGADM-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].COC(=O)CC1=CC=CC=C1N
1
–
1
von
1
Ergebnisse
1
D-(-)-2-Phenylglycin-Methylester-Hydrochlorid, 96 %, Thermo Scientific Chemicals
CAS: 19883-41-1 Summenformel: C9H12ClNO2 Molekulargewicht (g/mol): 201.65 MDL-Nummer: MFCD00137487 InChI-Schlüssel: DTHMTBUWTGVEFG-DDWIOCJRSA-N Synonym: h-d-phg-ome.hcl,h-d-phg-ome hcl,r-methyl 2-amino-2-phenylacetate hydrochloride,r-2-phenylglycine methyl ester hydrochloride,r---2-phenylglycine methyl ester hydrochloride,methyl 2r-amino phenyl acetate hydrochloride,d-alpha-phenylglycine methyl ester hydrochloride,d---2-phenylglycine methyl ester hydrochloride,methyl 2r-2-amino-2-phenylacetate hydrochloride,d-phenylglycine methyl ester hydrochloride PubChem CID: 12247453 IUPAC-Name: Methyl (2R)-2-amino-2-Phenylacetathydrochlorid SMILES: COC(=O)C(C1=CC=CC=C1)N.Cl