missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ zwf (Escherichia coli) Recombinant Protein

Used for Func, SDS-PAGE

Marke:  Abnova™ P4976.100ug

Additional Details : Gewicht : 0.02000kg

Artikelnummer. 16191700

  • 292.00 CHF / 100 Mikrogramm
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MAVTQTAQACDLVIFGAKGDLARRKLLPSLYQLEKAGQLNPDTRIIGVGRADWDKAAYTKVVREALETFMKETIDEGLWDTLSARLDFCNLDVNDTAAFSRLGAMLDQKNRITINYFAMPPSTFGAICKGLGEAKLNAKPARVVMEKPLGTSLATSQEINDQVGEYFEECQVYRIDHYLGKETVLNLLALRFANSLFVNNWDNRTIDHVEITVAEEVGIEGRWGYFDKAGQMRDMIQNHLLQILCMIAMSPPSDLSADSIRDEKVKVLKSLRRIDRSNVREKTVRGQYTAGFAQGKKVPGYLEEEGANKSSNTETFVAIRVDIDNWRWAGVPFYLRTGKRLPTKCSEVVVYFKTPELNLFKESWQDLPQNKLTIRLQPDEGVDIQVLNKVPGLDHKHNLQITKLDLSYSETFNQTHLADAYERLLLETMRGIQALFVRRDEVEEAWKWVDSITEAWAMDNDAPKPYQAGTWGPVASVAMITRDGRSWNEFE
Spezifikation

Spezifikation

NP_416366
Functional Study, SDS-PAGE
946370
zwf (Escherichia coli) Recombinant protein
Conventional Chromatography
100 ug
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
ECK1853/JW1841
zwf
E. coli
Escherichia coli expression system
>90% by SDS-PAGE
1 mg/mL
Liquid
27kDa
Escherichia coli expression system
15% SDS-PAGE Stained with Coomassie Blue
MAVTQTAQACDLVIFGAKGDLARRKLLPSLYQLEKAGQLNPDTRIIGVGRADWDKAAYTKVVREALETFMKETIDEGLWDTLSARLDFCNLDVNDTAAFSRLGAMLDQKNRITINYFAMPPSTFGAICKGLGEAKLNAKPARVVMEKPLGTSLATSQEINDQVGEYFEECQVYRIDHYLGKETVLNLLALRFANSLFVNNWDNRTIDHVEITVAEEVGIEGRWGYFDKAGQMRDMIQNHLLQILCMIAMSPPSDLSADSIRDEKVKVLKSLRRIDRSNVREKTVRGQYTAGFAQGKKVPGYLEEEGANKSSNTETFVAIRVDIDNWRWAGVPFYLRTGKRLPTKCSEVVVYFKTPELNLFKESWQDLPQNKLTIRLQPDEGVDIQVLNKVPGLDHKHNLQITKLDLSYSETFNQTHLADAYERLLLETMRGIQALFVRRDEVEEAWKWVDSITEAWAMDNDAPKPYQAGTWGPVASVAMITRDGRSWNEFE
RUO
zwf
Specific activity is 8-10 units/mL obtained by measuring the increase of NADPH in absorbance at 340nm resulting from the reduction of NAD or NADP. One unit oxidizes 1.0μmole D-glucose-6-phosphate to 6-phospho-D-gluconate per min in the presence of beta-
None
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote