missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZO-1 Mouse anti-Canine, Human, Mouse, Clone: 1G4A1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66452-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.Spezifikation
ZO-1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O97758, P39447, Q07157 | |
TJP1 | |
ZO-1 Fusion Protein Ag16454 | |
150 μL | |
Primary | |
Canine, Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1G4A1 | |
Unconjugated | |
TJP1 | |
Tight junction protein 1, Tight junction protein ZO 1, TJP1, ZO 1, ZO1, ZO-1, Zona occludens protein 1, Zonula occludens protein 1 | |
Mouse | |
Protein G | |
RUO | |
21872, 403752, 7082 | |
-20°C | |
Liquid |