missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF699 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21562-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ZNF699 may be involved in transcriptional regulation.Spezifikation
ZNF699 | |
Polyclonal | |
Unconjugated | |
ZNF699 | |
hang, Hangover homolog, zinc finger protein 699, ZNF699 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
374879 | |
Antibody | |
IgG |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q32M78 | |
ZNF699 | |
ZNF699 Fusion Protein Ag15990 | |
150 μL | |
Primary | |
Human | |
Liquid |