missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF557 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20500-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ZNF557 is a new candidate transcription factor.Spezifikation
ZNF557 | |
Polyclonal | |
Unconjugated | |
ZNF557 | |
zinc finger protein 557, ZNF557 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79230 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N988 | |
ZNF557 | |
ZNF557 Fusion Protein Ag14239 | |
150 μL | |
Primary | |
Human | |
Liquid |