missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF496 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20900-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ZNF496 contains a SCAN box, a KRAB-A domain, and four consensus C2H2-type zinc fingers preceded by a unique finger derivative, referred to herein as the C2HR motif. The C2HR motif functions to mediate protein-protein interaction with the cysteine-rich (C5HCH) domain of NSD1 in a Zn(II)-dependent fashion, and when tethered to RNA polymerase II promoters, represses transcription in an NSD1-dependent manner. ZNF496 contains a novel type of zinc finger motif that functions as a docking site for NSD1 and is more than just a degenerate evolutionary remt of a C2H2 motif.Spezifikation
ZNF496 | |
Polyclonal | |
Unconjugated | |
ZNF496 | |
NIZP1, zinc finger protein 496, ZKSCAN17, ZNF496 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
268417, 84838 | |
Antibody | |
IgG |
Western Blot | |
0.63 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5SXI5, Q96IT1 | |
ZNF496 | |
ZNF496 Fusion Protein Ag15015 | |
150 μL | |
Primary | |
Human, Mouse | |
Liquid |