missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF395 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11759-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Papillomavirus Regulatory Factor 1/HDBP2 is a novel transcription factor shuttling between nucleus and cytoplasm and binds to the specific GCCGGCG, which is an essential cis-element for Huntington′s disease gene expression.Spezifikation
ZNF395 | |
Polyclonal | |
Unconjugated | |
ZNF395 | |
HD regulating factor 2, HDBP 2, HDBP2, HDRF 2, Papillomavirus binding factor, PBF, PRF 1, PRF1, Si 1 8 14, zinc finger protein 395, ZNF395 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55893 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H8N7 | |
ZNF395 | |
ZNF395 Fusion Protein Ag2355 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |