Learn More
ZNF280C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563181
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PLQSKLPTAP FGCAPGTSFL QVTPPTSQNT TARNPRKSNA SRSKTSKLHA TTSTASKVNT SKPRGRIAKS KAKPSY Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 38%.
May function as a transcription factor.Spezifikation
ZNF280C | |
Polyclonal | |
Unconjugated | |
ZNF280C | |
SUHW3; Suppressor of hairy wing homolog 3; zinc finger protein 280C; Zinc finger protein 633; ZNF280C; ZNF633 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55609 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8ND82 | |
ZNF280C | |
Recombinant protein corresponding to Human ZNF280C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only