missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19429-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Dual bands of 50 kDa and 56 kDa detected by this antibody may represent the native and phosphorylated forms of ZIP7, respectively.
The zinc transporter ZIP7, also known as SLC39A7, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP7 was initially identified while characterizing genes in the major histocompatibility complex on chromosome 17. ZIP7 mRNA is abundantly and widely expressed and the protein localizes to the Golgi apparatus. It functions to transport intracellular zinc from the Golgi apparatus to the cytoplasm of the cell. ZIP7 expression is expressed by zinc. ZIP7 has been suggested to act a hub for tyrosine kinase activation and may thus be a potential therapeutic target for diseases such as cancer where prevention of tyrosine kinase activation would be advantageous.Spezifikation
ZIP7 | |
Polyclonal | |
Unconjugated | |
SLC39A7 | |
KE4, RING5, SLC39A7, Zinc transporter SLC39A7, ZIP 7, ZIP7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14977, 294281, 7922 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q31125, Q92504 | |
SLC39A7 | |
ZIP7 Fusion Protein Ag13762 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |