Learn More
Chemsplash Zip Labcoat T5/6 Fabric, Elastic Cuff
GREENER_CHOICE
Artikelnummer. 30058255
Click to view available options
Größe:
4X-Large
5X-Large
Packungsgröße:
50 Stück
Artikelnummer. 30058255
Marke: Chemsplash 2772.400.011
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Chemsplash Lab Coat is made from Chemsplash fabric which is soft and microporous. This lab coat is a 2/3 length coat with a collar and zip closure and elasticated wrists.
Chemsplash Lab Coat
  • 55GSM Type PB 6B Fabric
  • Zip Fasten Front with Adhesive Flap Cover
  • Elasticated Cuffs
  • Fold Over Collar
  • Anti-Static Material
  • Non-Linting
  • Silicone and Latex Free
  • PFAS Free
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Lab Coat is made from Chemsplash fabric, which is soft, microporous, and resistant to chemical splash and dust. This lab coat features a 2/3 length design with a collar and zip closure, along with elasticated wrists for a snug fit. It is anti-static to meet EN1149-5:2008 standards, lint-free, and suitable for cleanroom applications. The lab coat conforms to ISO 14644-1:2015 Cleanroom Class 5-9, ensuring it provides the protection required for sensitive environments. Chemsplash accessories are designed to protect both the wearer and the environment, offering maximum comfort and wearability.
Spezifikation
Produkttyp Lab Coat
Größe 5X-Large
Geschlecht Unisex
Material Microporous Composite Fabric
Farbe White
Verschlusstyp Zipper
Stulpenform Elastic
Einwegprodukt Single-use
Sterilität Non-sterile
Widerstand Liquids
Anzahl Taschen None
Latexfrei Yes
Autoklavierbar Autoclavable
Cleanroom Class Class 100-100,000 (ISO 5-8)
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato