Learn More
Chemsplash Zip Labcoat T5/6 Fabric, Elastic Cuff
Beschreibung
Chemsplash Lab Coat
- 55GSM Type PB 6B Fabric
- Zip Fasten Front with Adhesive Flap Cover
- Elasticated Cuffs
- Fold Over Collar
- Anti-Static Material
- Non-Linting
- Silicone and Latex Free
- PFAS Free
- Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Lab Coat is made from Chemsplash fabric, which is soft, microporous, and resistant to chemical splash and dust. This lab coat features a 2/3 length design with a collar and zip closure, along with elasticated wrists for a snug fit. It is anti-static to meet EN1149-5:2008 standards, lint-free, and suitable for cleanroom applications. The lab coat conforms to ISO 14644-1:2015 Cleanroom Class 5-9, ensuring it provides the protection required for sensitive environments. Chemsplash accessories are designed to protect both the wearer and the environment, offering maximum comfort and wearability.
Spezifikation
Spezifikation
| Produkttyp | Lab Coat |
| Größe | 5X-Large |
| Geschlecht | Unisex |
| Material | Microporous Composite Fabric |
| Farbe | White |
| Verschlusstyp | Zipper |
| Stulpenform | Elastic |
| Einwegprodukt | Single-use |
| Sterilität | Non-sterile |
| Widerstand | Liquids |
| Vedi altri risultati |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.