missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP36L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19005-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
TIS11D, also known as ZFP36LA (zinc finger protein 36, C3H type-like 2), BRF2 (butyrate response factor 2), ERF2 or RNF162C, is a 494 amino acid protein that localizes to the nucleus and contains two CSH1-type zinc fingers. Belonging to the TIS11 family of early response proteins, TIS11D is thought to function as a nuclear transcription factor that binds to 5′UUAUUUAUUU-3′ core RNA sequences and may regulate growth factor-induced cellular responses. The gene encoding TIS11D maps to human chromosome 2, which houses over 1, 400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes.Spezifikation
ZFP36L2 | |
Polyclonal | |
Unconjugated | |
ZFP36L2 | |
BRF2, Butyrate response factor 2, EGF response factor 2, ERF 2, ERF2, Protein TIS11D, RNF162C, TIS11D, ZFP36L2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12193, 298765, 678 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P23949, P47974 | |
ZFP36L2 | |
ZFP36L2 Fusion Protein Ag13518 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |