missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YME1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11510-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
YMEL1 is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies.Spezifikation
YME1L1 | |
Polyclonal | |
Unconjugated | |
YME1L1 | |
FTSH, FTSH1, Meg 4, MEG4, PAMP, YME1 like 1 (S. cerevisiae), YME1 like protein 1, YME1L, YME1L1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10730, 114217, 27377 | |
Antibody | |
IgG |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88967, Q925S8, Q96TA2 | |
Yme1l1 | |
YME1L1 Fusion Protein Ag2069 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Liquid |