missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XRCC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA576068
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad51. XRCC4 is an accessory factor of DNA ligase IV that preferentially binds DNA with nicks or broken ends. XRCC4 binds to DNA ligase IV and enhances its joining activity, and it is also involved in V(D)J recombination. Any defect in one of the known components of the DNA repair/V(D)J recombination machinery (Ku-70, Ku-80, DNA-PKCS, XRCC4 and DNA ligase IV) leads to abortion of the V(D)J rearrangement process and early block in both T and B cell maturation.
Spezifikation
| XRCC4 | |
| Polyclonal | |
| Unconjugated | |
| XRCC4 | |
| 2310057B22Rik; AW413319; AW545101; DNA repair protein XRCC4; SSMED; X-ray repair complementing defective repair in Chinese hamster cells 4; X-ray repair cross complementing 4; X-ray repair cross complementing protein 4; X-ray repair cross-complementing protein 4; XRCC4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 108138, 309995, 7518 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13426, Q924T3 | |
| XRCC4 | |
| Recombinant full length Human XRCC4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur