missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12830-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This locus controls the synthesis of the Kell blood group ′precursor substance′ (Kx). Mutations in this gene have been associated with McLeod syndrome, an X-linked, recessive disorder characterized by abnormalities in the neuromuscular and hematopoietic systems. XK has structural characteristics of prokaryotic and eukaryotic membrane transport proteins. This locus controls the synthesis of the Kell blood group ′precursor substance′ (Kx). Mutations in this gene have been associated with McLeod syndrome, an X-linked, recessive disorder characterized by abnormalities in the neuromuscular and hematopoietic systems. The encoded protein has structural characteristics of prokaryotic and eukaryotic membrane transport proteins.Spezifikation
XK | |
Polyclonal | |
Unconjugated | |
XK | |
Kell complex 37 kDa component, KX, Kx antigen, Membrane transport protein XK, X1k, XK, XK related protein 1, XKR1, XRG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22439, 497078, 7504 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51811, Q5GH61, Q9QXY7 | |
Xk | |
XK Fusion Protein Ag3550 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |