Learn More
Melford X-GlcA Cyclohexylammonium Salt
Click to view available options
Menge:
100 mg
250 mg
500 mg
1 g
Packungsgröße:
1 Gramm
100 Milligramm
250 Milligramm
500 Milligramm
Beschreibung
Histochemical substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene.
Histochemical substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene. The substrate is used as a qualitative histochemical marker of specific GUS expression in cells and tissue. X-GlcA is cleaved by GUS at the B1 glucuronic bond between glucuronic acid and the 5-Bromo-4-chloro-3-indolyl part of X-GlcA via hydrolysis. The cleavage of X-GlcA results in the precipitation of water insoluble blue dichloro-dibromo-indigo precipitate at the site of enzymatic cleavage. Allow to warm to room temperature before using.
Spezifikation
Spezifikation
| Menge | 500 mg |