missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford X-GAL
Click to view available options
Menge:
100 mg
250 mg
1 g
5 g
10 g
100 g
Packungsgröße:
1 Gramm
10 Gramm
100 Gramm
100 Milligramm
250 Milligramm
5 Gramm
Beschreibung
Chromogenic substrate of β-Galactosidase used in combination with IPTG for detection of β-Galactosidase activity in bacterial colonies.
Chromogenic substrate of β-Galactosidase used in combination with IPTG for detection of β-Galactosidase activity in bacterial colonies. X-GAL is cleaved at the B1-4 bond between galactose and the 5-Bromo-4-chloro-3-indolyl portion of X-GAL by β-Galactosidase via hydrolysis. The cleavage of X-GAL results in the production of a water insoluble blue dichloro-dibromo-indigo precipitate. In cloning strategies with vectors such as Lambda-11, M13mp18 and 19, pUC18 and 19, pUR222 the E. coli lacZ gene is transformed to lac+ cells. After transformation the cells show β-Galactosidase activity in the presence of media containing X-GAL and IPTG. The insertion of a DNA fragment into the cloning sites of the lacZ gene results in the disruption of β-Galactosidase activity leading to the appearance of white colonies. Non-recombinant cells produce a blue indigo dye.
Water Content: 0.05% (K. Fisher)
Protect from light and moisture
Spezifikation
Spezifikation
| Menge | 1 g |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur