missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12609-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Wilm′s Tumor (WT), a sporadic and familial childhood kidney tumor, is genetically heterogeneous. Wilm′s tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelioma.Spezifikation
WT1 | |
Polyclonal | |
Unconjugated | |
WT1 | |
GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT 2, WT1, WT33 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22431, 24883, 7490 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P19544, P22561, P49952 | |
WT1 | |
WT1 Fusion Protein Ag3337 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |