missing translation for 'onlineSavingsMsg'
Learn More

WFS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 11558-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838723

  • 131.19 CHF / 20 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

WFS1 is a novel component of Wolfram syndrome, a rare form of juvenile diabetes. WFS1 plays an important role in maintaining homeostasis of the endoplasmic reticulum (ER) in the pancreas. It is normally up-regulated during insulin secretion, whereas inactivation of the protein can cause ER stress. Chronic ER stress is a major involvement in Wolfram syndrome.
Spezifikation

Spezifikation

WFS1
Polyclonal
Unconjugated
WFS1
WFRS, WFS, WFS1, Wolfram syndrome 1 (wolframin), WOLFRAMIN
Rabbit
Antigen Affinity Chromatography
RUO
22393, 7466
Antibody
IgG
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O76024, P56695
WFS1
WFS1 Fusion Protein Ag2114
20 μL
Primary
Human, Mouse
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote