missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR43 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525402
Additional Details : Gewicht : 0.01000kg
Beschreibung
The function of WDR43 remains unknown.Spezifikation
WDR43 | |
Polyclonal | |
Unconjugated | |
WDR43 | |
2610318G08Rik; AU020887; AV024208; Kiaa0007; mKIAA0007; NET12; U3 small nucleolar RNA-associated protein 5 homolog; UTP5; UTP5, small subunit (SSU) processome component, homolog; WD repeat domain 43; WD repeat-containing protein 43; WDR43; Wrd43 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
23160 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q15061 | |
WDR43 | |
KLH conjugated synthetic peptide between 636-665 amino acids from the C-terminal region of human WDR43 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |