Learn More
Keystone Safety Wall Mountable Hair Net Dispenser
Artikelnummer. 17853532 Alle ansehen Keystone Cleanroom Products Produkte
Ansicht ändern
Click to view available options
Dispenser; Color: Clear; Size: Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
17853532 Dispenser; Color: Clear; Size: Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17853532 Lieferant Keystone Safety Lieferanten-Nr. 19055400
  • ONLINE EXCLUSIVE PRICE
    115.00 CHF
    90.46 CHF / 1 Stück
    Sparen 24.54 CHF
    21% Rabatt
 Eingeschränkter Artikel
Lager vorrätig
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Reduce wastage of hairnets, bouffant and beard covers with this functional dispenser. Constructed from sturdy and durable High Impact PETG, making it 19 times stronger than regular acrylic. Holds 100 hair nets/bouffant or 300 beard covers inside it and is wall mountable to better utilize your space.
  • PETG Material
  • Wall Mountable
  • 12 x 5 x 12 in.
  • Holds 100 pieces of hairnets/bouffant or 300 pieces of beard covers
Spezifikation
Produkttyp Dispenser
Farbe Clear
Tiefe (angloamerikanisch) 12 in.
Abmessungen (T x B x H) innen 12 x 5 x 12 in.
Breite (angloamerikanisch) 5 in.
Höhe (angloamerikanisch) 12 in.
Aufnahmevermögen PPE
Material PETG
Mounting Wall Mount
No. of Compartments 1
Product Style Hinged Lid with Round Front Opening
Zur Verwendung mit (Anwendung) Hold hairnets, bouffant, or beard covers
Größe Groß